Learn More
Invitrogen™ ADPRHL2 Polyclonal Antibody

Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
Specifications
Specifications
| Antigen | ADPRHL2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ADPRHL2 |
| Gene Accession No. | Q9NX46 |
| Gene Alias | [Protein ADP-ribosylarginine] hydrolase-like protein 2; [Protein ADP-ribosylserine] hydrolase; ADPRHL2; ADP-ribose glycohydrolase ARH3; ADP-ribose glycohydrolase ARH3; poly(ADP-ribose) glycohydrolase ARH3; ADP-ribosylarginine hydrolase like 2; ADP-ribosylhydrolase 2; ADP-ribosylhydrolase 3; ADP-ribosylhydrolase like 2; ADP-ribosylserine hydrolase; ADPRS; AI836109; ARH3; ecto ADP-ribosylhydrolase; LOW QUALITY PROTEIN: poly(ADP-ribose) glycohydrolase ARH3; O-acetyl-ADP-ribose deacetylase ARH3; Poly(ADP-ribose) glycohydrolase ARH3; protein ADP-ribosylarginine hydrolase-like protein 2 |
| Gene Symbols | ADPRHL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.