Learn More
ADRA1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is specific to ADRA1B.
Specifications
Specifications
| Antigen | ADRA1B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.24 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | ADRA1B |
| Gene Accession No. | P15823, P35368, P97717 |
| Gene Alias | ADRA1, ADRA1B, Alpha 1B adrenergic receptor, Alpha 1B adrenoceptor, Alpha 1B adrenoreceptor, ALPHA1BAR |
| Gene Symbols | Adra1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.