Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenalone Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. A123425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A123425G 25 g
A12345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A123425G Supplier TCI America Supplier No. A123425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62-13-5
Molecular Formula C9H12ClNO3
Molecular Weight (g/mol) 217.649
MDL Number MFCD00035075
InChI Key CSRRBDMYOUQTCO-UHFFFAOYSA-N
Synonym adrenalone hydrochloride, stryphnonasal, adrenone hydrochloride, adrenalone hcl, kephrine hydrochloride, adrenalonium chloratum, epinephrine ketone hydrochloride, 3',4'-dihydroxy-2-methylamino acetophenone hydrochloride, unii-nn82ywe2ic
PubChem CID 66136
IUPAC Name 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanone;hydrochloride
SMILES CNCC(=O)C1=CC(=C(C=C1)O)O.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 217.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Adrenalone Hydrochloride Hydrate
RTECSNumber AM8225000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.