Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Adrenocorticotropic Hormone (1-39), human

Catalog No. p-7076678
Click to view available options
Quantity:
0.5 mg
1 mg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12279-41-3

Specifications

Content And Storage Store at -20°C
Molecular Formula C207H308N56O58S
Format Solid
InChI Key IDLFZVILOHSSID-UHFFFAOYNA-N
SMILES CSCCC(NC(=O)C(CO)NC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C(N)CO)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CNC=N1)C(=O)NC(CC1=CC=CC=C1)C(=O)NC(CCCNC(N)=N)C(=O)NC(CC1=CNC2=CC=CC=C12)C(=O)NCC(=O)NC(CCCCN)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NCC(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NC(CCCCN)C(=O)NC(C(C)C)C(=O)NC(CC1=CC=C(O)C=C1)C(=O)N1CCCC1C(=O)NC(CC(N)=O)C(=O)NCC(=O)NC(C)C(=O)NC(CCC(O)=O)C(=O)NC(CC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CO)C(=O)NC(C)C(=O)NC(CCC(O)=O)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)N1CCCC1C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 4-({1-[(1-{[1-({1-[({[6-amino-1-(2-{[1-({[(5-amino-1-{[5-amino-1-({1-[(1-{2-[(1-{[5-amino-1-({1-[(1-{2-[(2-carbamoyl-1-{[({1-[(3-carboxy-1-{[2-carboxy-1-({3-carboxy-1-[(1-{[1-({3-carboxy-1-[(1-{[1-(2-{[1-({3-carboxy-1-[(1-carboxy-2-phenylethyl)carbamoyl]propyl}carbamoyl)-3-methylbutyl]carbamoyl}pyrrolidin-1-yl)-1-oxo-3-phenylpropan-2-yl]carbamoyl}ethyl)carbamoyl]propyl}carbamoyl)ethyl]carbamoyl}-2-hydroxyethyl)carbamoyl]propyl}carbamoyl)ethyl]carbamoyl}propyl)carbamoyl]ethyl}carbamoyl)methyl]carbamoyl}ethyl)carbamoyl]pyrrolidin-1-yl}-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamoyl]-2-methylpropyl}carbamoyl)pentyl]carbamoyl}-2-methylpropyl)carbamoyl]pyrrolidin-1-yl}-5-carbamimidamido-1-oxopentan-2-yl)carbamoyl]-4-carbamimidamidobutyl}carbamoyl)pentyl]carbamoyl}pentyl)carbamoyl]methyl}carbamoyl)-2-methylpropyl]carbamoyl}pyrrolidin-1-yl)-1-oxohexan-2-yl]carbamoyl}methyl)carbamoyl]-2-(1H-indol-3-yl)ethyl}carbamoyl)-4-carbamimidamidobutyl]carbamoyl}-2-phenylethyl)carbamoyl]-2-(1H-imidazol-4-yl)ethyl}carbamoyl)-4-(2-{2-[2-(2-amino-3-hydroxypropanamido)-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido}-4-(methylsulfanyl)butanamido)butanoic acid
Molecular Weight (g/mol) 4541.14
Quantity 0.5 mg

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.