Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adrenomedullin/ADM Antibody (OTI8A1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270100AF750
Description
Adrenomedullin/ADM Monoclonal specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry.Specifications
Adrenomedullin/ADM | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ADM | |
Recombinant protein fragment corresponding to amino acids 22-146 of human Adrenomedullin/ADM (NP_001115) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Immunohistochemistry | |
OTI8A1 | |
Immunohistochemistry | |
adrenomedullin, AMpreproadrenomedullin | |
Mouse | |
Affinity Purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
133.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction