Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adrenomedullin Monoclonal Antibody (1D2C2C10)
GREENER_CHOICE

Catalog No. ABS0270102
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0270102 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0270102 Supplier Invitrogen™ Supplier No. ABS0270102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The epitope of this antibody has been mapped to a region within residues 22-52. This antibody shows no cross-reactivity to CGRP. In ELISA applications, the binding of this antibody is not inhibited by full-length adrenomedullin in solution up to a concentration of 1 μmol/L, but is inhibited by adrenomedullin (aa 22-52) in solution. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adrenomedullin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D2C2C10
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene ADM
Gene Accession No. P35318
Gene Alias ADM; Adrenomedullin; AM; AMpreproadrenomedullin; Ap; H39316; PAMP; preproadrenomedullin; proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide; ProAM-N20; RATAP
Gene Symbols ADM
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues 1-52 of human adrenomedullin.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.