Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adrenomedullin R/ADMR/GPR182 Antibody (528563) [DyLight 350], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10293D
Description
Adrenomedullin R/ADMR/GPR182 Monoclonal antibody specifically detects Adrenomedullin R/ADMR/GPR182 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Adrenomedullin R/ADMR/GPR182 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
528563 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
ADMR, adrenomedullin receptor, AMR, AM-R, G protein-coupled receptor 182,7TMR, G10D, gamrh, G-protein coupled receptor 182, hrhAMR, MGC34399 | |
HEK293 human embryonic kidney cell line transfected with human Adrenomedullin R/ADMR, Accession # O15218 | |
0.1 mL | |
GPCR | |
11318 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction