Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADRM1 Antibody (34C2), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259402PCP
Description
ADRM1 Monoclonal antibody specifically detects ADRM1 in Human samples. It is validated for Flow CytometrySpecifications
ADRM1 | |
Monoclonal | |
PerCP | |
000 surface antigen, 110 kDa cell membrane glycoprotein, adhesion regulating molecule 1, Adhesion-regulating molecule 1, ARM1, ARM-1, Gp110, GP110hRpn13, M(r) 110, MGC29536, proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, regulatory particle non-ATPase 13, Rpn13, Rpn13 homolog | |
Recombinant human ADRM1 (1-407aa) purified from E. coli | |
0.1 mL | |
Cancer | |
11047 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry | |
34C2 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction