Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADRM1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130446F
Description
ADRM1 Polyclonal antibody specifically detects ADRM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ADRM1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
000 surface antigen, 110 kDa cell membrane glycoprotein, adhesion regulating molecule 1, Adhesion-regulating molecule 1, ARM1, ARM-1, Gp110, GP110hRpn13, M(r) 110, MGC29536, proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, regulatory particle non-ATPase 13, Rpn13, Rpn13 homolog | |
The immunogen for this product maps to a region between residue 300 and 350 of human adhesion regulating molecule 1 using the numbering given in entry NP_008933.2 (GeneID 11047). | |
0.1 mL | |
Cancer | |
11047 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction