Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559930 Supplier Invitrogen™ Supplier No. PA559930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATMNVPAGPA GGQQVDLASV LTPEIMAPIL ANADVQERLL PYLPSGESLP QTADEIQNTL TSPQFQQALG MFSAALASGQ LGPLMCQFGL PAEAVEAANK GDVEAFAKAM Q Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADRM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADRM1
Gene Accession No. Q16186
Gene Alias 110 kDa cell membrane glycoprotein; 1110063P18Rik; 2510006J17Rik; AA408205; adhesion regulating molecule 1; adhesion-regulating molecule 1; ADRM1; Arm1; ARM-1; AU043535; cell membrane glycoprotein, 110000M(r) (surface antigen); glycoprotein 110; GP110; hRpn13; M(r) 110,000 surface antigen; MGC29536; proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13; rpn13 homolog
Gene Symbols ADRM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADRM1. Recombinant protein control fragment (Product #RP-89612).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11047
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.