Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adropin Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126387AF405
Description
Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.Specifications
Adropin | |
Polyclonal | |
Western Blot | |
Q6UWT2 | |
ENHO | |
Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-Prot# Q6UWT2] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. 88% sequence identity with bovine protein. | |
Store at 4C in the dark. |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 | |
Rabbit | |
8 kDa | |
0.1 mL | |
Diabetes Research, Lipid and Metabolism | |
375704 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction