Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adropin Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126387AF532
Description
Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.Specifications
Adropin | |
Polyclonal | |
Western Blot | |
adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470 | |
Rabbit | |
Affinity Purified | |
Diabetes Research, Lipid and Metabolism | |
375704.0 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ENHO | |
Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-ProtNo. Q6UWT2] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction