Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adropin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572781
Description
Adropin Polyclonal Antibody for Western Blot, IHC (P), ELISA
Adropin is involved in the regulation of glucose homestasis and lipid metabolism. It is a secreted protein encoded by the Energy Homeostasis Associated gene (ENHO) found in the brain and liver. In the liver, the protein has been found to increase in response to high-fat foods and decrease after fasting. The protein seems to play a role in controlling the activity of other metabolic genes, specifically those involved in the production of lipids from carbohydrates.
Specifications
Adropin | |
Polyclonal | |
Unconjugated | |
ENHO | |
1110065P19Rik; 2310040A07Rik; Adropin; adropin-like; C9orf165; energy homeostasis associated; energy homeostasis-associated protein; ENHO; GAAI470; LOC100912292; RGD1565232; UNQ470; UNQ470/PRO830 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100912292, 375704, 69638 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6UWT2, Q8K1D8 | |
ENHO | |
ADROPIN antibody for an 18 amino acid peptide near the carboxy terminus of human ADROPIN. The immunogen is within the last 50 amino acids of ADROPIN. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction