Learn More
Invitrogen™ Adropin Polyclonal Antibody

Description
Suggested positive control: Adropin Overexpression lysate, mouse brain tissue lysate. 88% sequence identity with bovine protein.
Specifications
Specifications
| Antigen | Adropin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | ENHO |
| Gene Accession No. | Q6UWT2, Q8K1D8 |
| Gene Alias | 1110065P19Rik; 2310040A07Rik; Adropin; adropin-like; C9orf165; energy homeostasis associated; energy homeostasis-associated protein; ENHO; GAAI470; LOC100912292; RGD1565232; UNQ470; UNQ470/PRO830 |
| Gene Symbols | ENHO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.