Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adropin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989425
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9425 Supplier Bioss Supplier No. BS9886R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adropin is involved in the regulation of glucose homestasis and lipid metabolism. It is a secreted protein encoded by the Energy Homeostasis Associated gene (ENHO) found in the brain and liver. In the liver, the protein has been found to increase in response to high-fat foods and decrease after fasting. The protein seems to play a role in controlling the activity of other metabolic genes, specifically those involved in the production of lipids from carbohydrates.

Specifications

Antigen Adropin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ENHO
Gene Accession No. Q6UWT2
Gene Alias 1110065P19Rik; 2310040A07Rik; Adropin; adropin-like; C9orf165; energy homeostasis associated; Energy homeostasis-associated protein; Enho; GAAI470; LOC100912292; RGD1565232; UNQ470; UNQ470/PRO830
Gene Symbols ENHO
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Adropin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375704
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.