Learn More
ADRP/Perilipin 2 Mouse anti-Human, Clone: 1G5D4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ADRP/Perilipin 2 |
---|---|
Clone | 1G5D4 |
Concentration | 1.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6427
![]() ![]() |
Proteintech Group Inc
603401IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Anti-ADRP antibody is a reliable and sensitive marker for lipid droplets. Enhanced expression of ADRP is linked to diseases with abnormal lipid storage, including hepatic steatosis, atherosclerosis and diabetes. Immunohistochemistry of ADRP may facilitate
Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.Specifications
ADRP/Perilipin 2 | |
1.17 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ADFP, Adipophilin, ADRP, Perilipin 2, PLIN2 | |
PLIN2 | |
IgG1 | |
Protein G | |
Antibody |
1G5D4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q99541 | |
123 | |
ADRP/Perilipin 2 Fusion Protein Ag7643 | |
Primary | |
-20°C | |
PLIN2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.