Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADSL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580324
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ADSL/Adenylosuccinate Lyase.
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.
Specifications
ADSL | |
Polyclonal | |
Unconjugated | |
Adsl | |
2510006M18Rik; Adenylosuccinase; adenylosuccinate lyase; adenylosuccinate lyase 1; Adl; Adsl; AMPS; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; ASase; ASL; null; test; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P30566 | |
Adsl | |
Recombinant Human ADSL/Adenylosuccinate Lyase protein (Met1-Leu484). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction