Learn More
Invitrogen™ ADSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529964
Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (95%), Zebrafish (84%), Xenopus laevis (90%), Pig (97%), Chicken (91%), Chimpanzee (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.
Specifications
| ADSL | |
| Polyclonal | |
| Unconjugated | |
| Adsl | |
| 2510006M18Rik; Adenylosuccinase; adenylosuccinate lyase; adenylosuccinate lyase 1; Adl; Adsl; AMPS; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; ASase; ASL; null; test; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11564, 158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P30566, P54822 | |
| Adsl | |
| Recombinant protein encompassing a sequence within the center region of human ADSL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.