Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Adson Forceps
SDP

Catalog No. p-7166911 Shop All Stoelting Co Products
Click to view available options
Length (English):
4.75 in.
7.25 in.
Model:
1 x 2 teeth
2 x 3 teeth
Curved
Serrarated teeth
Straight
TC, Cross serrated

Tissue forceps with the opposing blade surfaces covered with long, needle-like teeth.

Specifications

For Use With (Application) Neuroscience
Length (English) 7.25 in.
Model Straight
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.