Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADSS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555070
Description
Immunogen sequence: KLDGEIIPHI PANQEVLNKV EVQYKTLPGW NTDISNARAF KELPVNAQNY VRFIEDELQI PVKWIGVGKS RESM Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes the enzyme adenylosuccinate synthetase which catalyzes the first committed step in the conversion of inosine monophosphate to adenosine monophosphate. A pseudogene of this gene is found on chromosome 17.
Specifications
| ADSS | |
| Polyclonal | |
| Unconjugated | |
| ADSS2 | |
| ADEH; adenylosuccinate synthase; adenylosuccinate synthetase (Ade(-)H-complementing); adenylosuccinate synthetase 2, non muscle; adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; adenylosuccinate synthetase, liver isozyme; adenylosuccinate synthetase, non muscle; adenylosuccinate synthetase, non-muscle isozyme; Adss; ADSS 2; Adss2; AI314886; AMPSase 2; AS; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P30520 | |
| ADSS2 | |
| Recombinant protein corresponding to Human ADSS. Recombinant protein control fragment (Product #RP-93986). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction