Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADSS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109365 Supplier Invitrogen™ Supplier No. PA5109365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAFAETYPAA SSLPNGDCGR PRARPGGNRV TVVLGAQWGD EGKGKVVDLL AQDADIVCRC QGGNNAGHTV VVDSVEYDFH LLPSGIINPN VTAFIGNGVV IHLPGLFEEA EKNVQKGKGL EGWEKRLIIS DRAHIVFDFH QAADGIQEQQ RQEQAGKNLG TTKKGIGPVY SSKAARSGLR MCDLVSDFDG FSERFKVLAN QYKSIYPTLE IDIEGELQKL KGYMEKIKPM VRDGVYFLYE ALHGPPKKIL VEGANAALLD IDFGTYPFVT SSNCTVGGVC.

This gene encodes the enzyme adenylosuccinate synthetase which catalyzes the first committed step in the conversion of inosine monophosphate to adenosine monophosphate. A pseudogene of this gene is found on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADSS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADSS2
Gene Accession No. P30520, P46664
Gene Alias ADEH; adenylosuccinate synthase; adenylosuccinate synthetase (Ade(-)H-complementing); adenylosuccinate synthetase 2, non muscle; adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; adenylosuccinate synthetase, liver isozyme; adenylosuccinate synthetase, non muscle; adenylosuccinate synthetase, non-muscle isozyme; Adss; ADSS 2; Adss2; AI314886; AMPSase 2; AS; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase
Gene Symbols ADSS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADSS (NP_001117.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11566, 159
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.