Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114310 Supplier Invitrogen™ Supplier No. PA5114310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody specifically recognizes Arogenate dehydratase in Oryza sativa subspecies japonica (Rice).

Converts the prephenate produced from the shikimate-chorismate pathway into phenylalanine. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ADT2
Gene Accession No. Q9SSE7
Gene Alias ADT2; Arabidopsis thaliana arogenate dehydratase 2; arogenate dehydratase 2; Arogenate dehydratase/prephenate dehydratase 2, chloroplastic; At3g07630; AtADT2; MLP3.8; PDT2; putative P-protein; putative P-protein: chorismate mutase, prephenate dehydratase
Gene Symbols ADT2
Host Species Rabbit
Immunogen Synthetic peptide within rice Arogenate dehydratase aa 28-77.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 819954
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.