Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Advanced Crystal Violet Stain
SDP

Catalog No. p-200005336
Click to view available options
Volume (Metric):
250 mL
3.7 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a primary stain in the advanced gram stain procedure. An aqueous solution of crystal violet.

  • Crystal Violet is taken up equally well by both gram-positive and negative bacterial cell walls. A crystal violet-iodine complex is formed within the peptidoglycan layer of the cell wall of each type of bacteria upon addition of iodine
  • The advanced system helps to guard against accidental over-decolorization, thus reducing the possibility of mistaking a gram-positive organism for a gram-negative one

Specifications

Format Liquid
Color Violet
Volume (Metric) 250 mL
Compatible Cells Gram-negative and gram-positive bacteria
Label or Dye Dye
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.