Learn More
Advanced Instruments Drip Return Wiper
SDP
Catalog No. 22047107
Click to view available options
Packaging Quantity:
10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Sponges used to clean osmometer probes
Specifications
Product Type Drip Return Wipes
For Use With (Equipment) Model 3900 Osmometer, cleans osmometer probes
Packaging Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.