Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advanced Organic Stereochemistry Model Set
SCI_ED

Catalog No. s94024
Click to view available options
:
Organic stereochemistry model set

Includes

Kit Contents:

  • 12 black 4-peg carbon atom centers
  • 10 black 3-peg carbon atom centers
  • 3 red 4-peg atom centers
  • 3 blue 4-peg atom centers
  • 3 red 1-peg atom centers
  • 8 green 1-peg atom centers
  • 3 blue 1-peg atom centers
  • 18 white 1-peg hydrogen atom centers
  • 3 silver 6-peg atom centers
  • 3 purple 5-peg atom centers
  • 26-1.25 in. PVC, clear bonds (single bonds)
  • 2-0.82 in. PVC, clear bonds (sigma bonds)
  • 4-2 in. white bonds (pi bonds)
  • 10-1.25 in. grey, rigid bonds (rotating bonds)
  • Instructions

Expands understanding of stereochemistry with sp3, sp2, and sp bonding, aromatic rings, and trigonal planar atom centers.

This kit is an expanded version of the Organic Stereochemistry Model Set (S94023). This larger kit includes parts that allow the comparison of sp3, sp2, and sp bonding, and to make aromatic rings with trigonal planar atom centers.

  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing

Specifications

Product Type Model Set, Advanced Organic Stereochemistry
For Use With (Application) Understanding of stereochemistry with sp3, sp2 and sp bonding, aromatic rings and trigonal planar atom center
Includes 16 black 4-peg Carbon atom centers, 7 black 3-peg Carbon atom centers, 6 red 2-peg Oxygen atom centers, 37 white 1-peg Hydrogen atom centers, 2 green 1-peg Chlorine atom centers, 2 dark green 1-peg Bromine atom centers, 2 light green 1-peg Fluorine atom centers, 50-1.25 in. PVC, clear bonds (single bonds), 8-1.625 in. PVC, clear bonds (double bonds), lab procedure, student response form and answer key
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.