Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One Advanced TC Cell Culture Dishes
Shop All Greiner Bio-One Products

Click to view available options
Diameter (Metric):
100 mm
145 mm
Volume (Metric) Working:
15 to 17 mL
25 to 27 mL
Height (Metric):
20 mm
Description
- Non-pyrogenic and non-cytotoxic
- Optimised cell adherence
- Increased cell yield
- Homogenous cell growth
- Higher transgene activity
- Improved assay consistency
- Non-biological surface, sterile
- Shelf
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 240 mL |
| Color | Clear |
| Culture Area | 143 sq. cm |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction