Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One Advanced TC Treated Sterile Cell Culture Multi-well Plates
Shop All Greiner Bio-One Products
Click to view available options
Well Count:
12
24
48
Capacity (Metric):
1.7 mL
3.3 mL
6.5 mL
Description
- Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
- For use with serum reduced and serum-free media
- Allows differentiation of semi-adherent cells
- Non-pyrogenic and non-cytotoxic
- Lid with condensation rings
Warranty and Services
No
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 1.7 mL |
| Color | Clear |
| Culture Area | 1 sq. cm |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction