Learn More
Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Standard Screw Cap
GREENER_CHOICE
Catalog No. p-7075415
Click to view available options
Culture Area:
175 cm2
25 cm2
75 cm2
Volume (Metric) Working:
50mL
250mL
550 mL
650mL
For maintenance of adherent cells or suspension cell culture. Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Standard Screw Cap provides optimal conditions for cultivation of sensitive and fastidious cells.
  • Made of high-grade polystyrene
  • Free of detectable DNases, RNases, human DNA, endotoxins and are non-cytotoxic
  • Stacking rim on the top of the flasks ensures firm standing and easy stackability in the incubator
  • Printed graduation for easier filling on both sides
  • Provides homogeneous and optimized cell growth
  • Standard screw cap
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Surface Coating Advanced TC
Closure Type Standard Screw Cap
Culture Area 75 cm2
Volume (Metric) Working 250mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.