Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Advanced Thermostat Kit
SDPSCI_ED

Catalog No. p-7231594
Click to view available options
Quantity:
1 Ea.
5 Pk.

Observe energy transfer and conversion and investigate the systems that keep homes comfortable and safe with this advanced thermostat kit.

This apparatus helps students to observe energy transfer and conversion and investigate the systems that keep homes comfortable and safe. The suggested activities below will: explore the relationship between parallel circuits and current; understand how thermostats and circuit breakers operate; and compare the energy efficiency of LEDs to incandescent bulbs.

Components:
  • 6V DC Connections
  • Multimeter Connections
  • Bulb Sockets (5)
  • Bimetallic Bar Switch
  • On/off Switches (5)
  • 6V Incandescent Bulbs (5)
  • LED Bulbs (5)
  • Candle
  • Multimeter
  • Additional Supplies Needed: 6V DC Power Supply and Stopwatch

Specifications

Grade 6
Age 12 to 20
For Use With (Application) Teaching about Thermostats
Includes Advanced thermostat apparatus, 5 incandescent bulbs, 5 LED bulbs, Multimeter, Candle, Activity Guide
Quantity 1 Ea.
Product Type Physics Demonstration
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.