Learn More
Specifications
Specifications
| Certifications/Compliance | NIOSH, EN |
| Connection Type | Dual-canister mount (bayonet mount) |
| Material | Polycarbonate Wrap-around Lens |
| Product Type | Advantage™ 3200 Gas Mask |
| Format | Small |
| For Use With (Application) | Industrial applications |
| Includes | Advantage™ head harness, canister, nosecup and identification tag |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.