Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 3200 CBA-RCA Gas Mask

Catalog No. 19121718B Shop All MSA Products
Change view
Click to view available options
Includes:
Advantage™ head harness, canister, nosecup and identification tag
Rubber head harness, canister, nosecup and identification tag
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-121-718B Advantage™ head harness, canister, nosecup and identification tag
19-121-719C Rubber head harness, canister, nosecup and identification tag
19-121-718C Advantage™ head harness, canister, nosecup and identification tag
19-121-718A Advantage™ head harness, canister, nosecup and identification tag
19-121-719A Rubber head harness, canister, nosecup and identification tag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-121-718B Supplier MSA™ Supplier No. 10036325
This item is not returnable. View return policy

Specifications

Certifications/Compliance NIOSH, EN
Connection Type Dual-canister mount (bayonet mount)
Material Polycarbonate Wrap-around Lens
Product Type Advantage™ 3200 Gas Mask
Format Medium
For Use With (Application) Industrial applications
Includes Advantage™ head harness, canister, nosecup and identification tag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.