Description
- Approved with all MSA Advantage Cartridges
- Features two mounting locations for cartridges or filters on the right and left sides
- Standard nose cup with twin inhalation check valves
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirement.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1 2003, NIOSH 42 CFR Part 84 |
| Material | Silicone |
| Product Type | Twin-Port Facepiece |
| Size | Small |
| For Use With (Application) | Respiratory applications |
| For Use With (Equipment) | Msa™ respirator models |
| Harness Style | Dupont™ Kevlar™ aramid fiber net |
| Includes | Nosecup, bayonet adapters in lens, facepiece elastomers |
| Suspension Type | 4-point |