Learn More
MSA™ Advantage™ 4000 Twin-Port Facepiece
Catalog No. 17200090 Shop All MSA Products
Change view
Click to view available options
Material:
Rubber
Silicone
Silicone, Rubber
Size:
Small
Medium
Large
Harness Style:
Adjustable silicone rubber
Dupont™ Kevlar™ aramid fiber net
Kevlar net, four-point
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size Harness Style
17-200-090 Rubber Small Dupont™ Kevlar™ aramid fiber net
17-200-088 Rubber Medium Dupont™ Kevlar™ aramid fiber net
17-200-092 Rubber Large Dupont™ Kevlar™ aramid fiber net
17-200-089 Silicone Small Dupont™ Kevlar™ aramid fiber net
17-200-087 Silicone, Rubber Medium Kevlar net, four-point
17-200-091 Silicone Large Dupont™ Kevlar™ aramid fiber net
18-139-431 Rubber Medium Adjustable silicone rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-200-090 Supplier MSA™ Supplier No. 10108570
Add to Cart
Add to Cart
This item is not returnable. View return policy
Two bayonet cartridge adapters mount directly onto the facepiece lens for compatibility with all Advantage cartridges and filters
  • Approved with all MSA Advantage Cartridges
  • Features two mounting locations for cartridges or filters on the right and left sides
  • Standard nose cup with twin inhalation check valves
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirement.
Specifications
Certifications/Compliance ANSI Z87.1 2003, NIOSH 42 CFR Part 84
Material Rubber
Product Type Twin-Port Facepiece
Size Small
For Use With (Application) Respiratory applications
For Use With (Equipment) Msa™ respirator models
Harness Style Dupont™ Kevlar™ aramid fiber net
Includes Nosecup, bayonet adapters in lens, facepiece elastomers
Suspension Type 4-point