Learn More
MSA™ Advantage™ 900 Elastomeric Half-Mask Respirators
Catalog No. 19003388 Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-003-388 Small
19-003-389 Medium
19-003-390 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-003-388 Supplier MSA™ Supplier No. 10218527
Add to Cart
Add to Cart
An elastomeric half-mask respirator providing healthcare professionals, essential workers and first responders a secure protection solution with enhanced communication.
The Advantage 900 Elastomeric half-mask respirator provides healthcare professionals, essential workers and first responders a secure protection solution with enhanced communication.
Elastomeric respirators are designed for repeated cleaning and decontamination, resulting in dramatic cost savings over filtering facepiece respirators. The included mechanical speaking diaphragm allows for improved communication among essential workers, patients and the public.
  • No exhalation valve, allowing for filtration of exhaled air and providing a means of source control and use in a sterile field environment
  • Reuse and cleaning capability make for a more eco-friendly alternative
  • Overmolded MultiFlex nosecup provides stability on user's face while maintaining a secure fit
  • Anthrocurve facepiece design maximizes fit and comfort and reduces potential leak points
Specifications
Connection Type Bayonet
Color Blue
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
With Exhalation Valve No
Latex Free Yes
Sterility Non-sterile