Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AE Binding Protein 1/ACLP Antibody (590831), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6425F
Description
AE Binding Protein 1/ACLP Monoclonal antibody specifically detects AE Binding Protein 1/ACLP in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
AE Binding Protein 1/ACLP | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
165 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry (Intracellular Staining), CyTOF | |
590831 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ACLP, ACLPadipocyte enhancer binding protein 1, AE binding protein 1, AE-binding protein 1adipocyte enhancer-binding protein 1, Aortic carboxypeptidase-like protein, FLJ33612 | |
Mouse myeloma cell line NS0-derived recombinant human ACLP, Glu345-Phe1158, Accession # Q8IUX7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction