Learn More
Description
Specifications
Specifications
| Antigen | AE2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | P04920 |
| Gene Alias | AE 2, AE2anion exchanger 2 type a, Anion exchanger 2, anion exchanger 2 type b2, BND3LHKB3anion exchange protein 2, EPB3L1anion exchanger 2 type b1, FLJ59028, MPB3L, NBND3, Non-erythroid band 3-like protein, Solute carrier family 4 member 2, solute carrier family 4, anion exchanger, member 2 (erythrocyte membraneprotein band 3-like 1) |
| Gene Symbols | SLC4A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
