Learn More
Invitrogen™ AEBP1 Polyclonal Antibody

Description
AEBP1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | AEBP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | AEBP1 |
| Gene Accession No. | Q8IUX7 |
| Gene Alias | ACLP; adipocyte enhancer-binding protein 1; AE binding protein 1; AE-binding protein 1; AEBP1; Aortic carboxypeptidase-like protein; FLJ33612; transcription factor AEBP1 |
| Gene Symbols | AEBP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.