Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AEBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584677 Supplier Invitrogen™ Supplier No. PA584677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPVKPLLPPL PPDYGDGYVI PNYDDMDYYF GPPPPQKPDA ERQTDEEKEE LKKPKKEDSS PKEETDKWAV EKGKDH Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 77%.

The adipocyte enhancer binding protein 1 (AEBP1) is a transcriptional repressor with carboxypeptidase (CP) activity. This protein binds to a regulatory sequence, adipocyte enhancer 1 (AE-1), located in the proximal promoter region of the adipose P2 (aP2) gene, which encodes the adipocyte fatty-acid binding protein. It is characterized as a member of the regulatory B-like CP family. This protein seems to be activated by a novel mechanism, whereby the direct binding of DNA enhances its protease activity. AEBP1 may play a role in differentiated vascular smooth muscle cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AEBP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AEBP1
Gene Accession No. Q8IUX7
Gene Alias ACLP; adipocyte enhancer-binding protein 1; AE binding protein 1; AE-binding protein 1; AEBP1; Aortic carboxypeptidase-like protein; FLJ33612; transcription factor AEBP1
Gene Symbols AEBP1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human AEBP1 (Product #RP-103808)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.