Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AEBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111258
Description
Immunogen sequence: PPVKPLLPPL PPDYGDGYVI PNYDDMDYYF GPPPPQKPDA ERQTDEEKEE LKKPKKEDSS PKEETDKWAV EKGKDH.
The adipocyte enhancer binding protein 1 (AEBP1) is a transcriptional repressor with carboxypeptidase (CP) activity. This protein binds to a regulatory sequence, adipocyte enhancer 1 (AE-1), located in the proximal promoter region of the adipose P2 (aP2) gene, which encodes the adipocyte fatty-acid binding protein. It is characterized as a member of the regulatory B-like CP family. This protein seems to be activated by a novel mechanism, whereby the direct binding of DNA enhances its protease activity. AEBP1 may play a role in differentiated vascular smooth muscle cells.
Specifications
AEBP1 | |
Polyclonal | |
Unconjugated | |
AEBP1 | |
ACLP; adipocyte enhancer-binding protein 1; AE binding protein 1; AE-binding protein 1; AEBP1; Aortic carboxypeptidase-like protein; FLJ33612; transcription factor AEBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUX7 | |
AEBP1 | |
Recombinant Protein corresonding to Human AEBP1. Recombinant protein control fragment (Product #RP-103808). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction