Learn More
Invitrogen™ AEBP2 Monoclonal Antibody (3E3C10)

Description
MA5-31613 has been tested in indirect ELISA.
Specifications
Specifications
| Antigen | AEBP2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3E3C10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | AEBP2 |
| Gene Accession No. | Q6ZN18 |
| Gene Alias | adipocyte enhancer-binding protein 2; Adipocyte enhancer-binding protein 2 homolog; AE binding protein 2; AE(adipocyte enhancer)-binding protein 2; AE-binding protein 2; AE-binding protein 2 homolog; aebp2; AU023766; B230313N05Rik; LOW QUALITY PROTEIN: zinc finger protein AEBP2; si:dkey-158p11.2; wu:fk42f11; wu:fk52h12; zgc:63755; Zinc finger protein AEBP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.