Learn More
AEBP2 Mouse anti-Human, Porcine, Clone: 7H2B11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | AEBP2 |
---|---|
Clone | 7H2B11 |
Concentration | 0.93 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6576
![]() ![]() |
Proteintech Group Inc
661411IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody recognizes adult-specific larger form (52 kDa) and the embryo-specific smaller form (31 kDa).
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.Specifications
AEBP2 | |
0.93 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AE binding protein 2, AEBP2, Zinc finger protein AEBP2 | |
AEBP2 | |
IgG1 | |
Protein G | |
Antibody |
7H2B11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
Q6ZN18 | |
100522805, 121536 | |
AEBP2 Fusion Protein Ag17905 | |
Primary | |
-20°C | |
AEBP2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.