Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AEBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596666 Supplier Invitrogen™ Supplier No. PA596666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STISSGRSTP AMMNGQGSTT SSSKNIAYNC CWDQCQACFN SSPDLADHIR SIHVDGQRGG VFVCLWKGCK VYNTPSTSQS WLQRHMLTHS GDKPFKCVVG GCNASFASQG GLARHVPTHF SQQNSSKVSS QPKAKEESPS KAGMNKRRKL KNKRRRSLPR PHDFFDAQTL DAIRHRAICF NLSAHIESLG KGHSVVFHST VIAKRKEDSG KIKLLLHWMP EDILPDVWVN ESERHQLKTK VVHLSKLPKD TALLLDPNIY RTMPQKRLKR; Positive Samples: Jurkat, 293T, A-549, Mouse kidney, Mouse skeletal muscle, Rat heart; Cellular Location: Nucleus.

AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates 'Lys-9' and 'Lys-27' residues of histone H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AEBP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AEBP2
Gene Accession No. Q6ZN18, Q9Z248
Gene Alias adipocyte enhancer-binding protein 2; Adipocyte enhancer-binding protein 2 homolog; AE binding protein 2; AE(adipocyte enhancer)-binding protein 2; AE-binding protein 2; AE-binding protein 2 homolog; aebp2; AU023766; B230313N05Rik; LOW QUALITY PROTEIN: zinc finger protein AEBP2; si:dkey-158p11.2; wu:fk42f11; wu:fk52h12; zgc:63755; Zinc finger protein AEBP2
Gene Symbols AEBP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 234-503 of human AEBP2 (NP_6949392).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11569, 121536, 297705
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.