Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals AEBSF hydrochloride, 98%

Catalog No. p-50536
Click to view available options
Quantity:
50 mg
1 g
Packaging:
Glass bottle

CAS: 30827-99-7 | C8H10FNO2S·ClH | 239.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H10FNO2S·ClH
Molecular Weight (g/mol) 239.7
MDL Number MFCD00132962
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 175.0°C to 177.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 mg
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in absolute ethanol
Formula Weight 239.7
Percent Purity 98%
Physical Form Powder
Chemical Name or Material AEBSF hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.