Learn More
ZOLL™ Medical AED Plus™ Accessories
SCI_ED
Catalog No. p-7110765 Shop All Zoll Medical Corporation Products
Click to view available options
Product Type:
Cabinet, Standard Metal Wall
Carry Bag
Pelican Case
Wall Mounting Bracket
Catalog No. S38755
Supplier: ZOLL™ Medical 8000083701/EMD
Accessories for AED Plus Defibrillator
  • Large waterproof hard-shell carry case that stores the AED Plus, one set of CPR-D-padz™, one set of Pedi-padz™ II and a sleeve of batteries
  • High-density foam provides total protection from the rigors of impact, vibration, and shock
Specifications
Product Type Pelican Case
Size Large
Includes Case that holds AED Plus, CPR-D·padz, and pedi·padz II
For Use With (Equipment) AED Plus Defibrillator
Certifications/Compliance IP67, C-4150J
Max. Temperature (English) 210°F
Max. Temperature (Metric) 99°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.