Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AEE 788
GREENER_CHOICESDP

Catalog No. 531850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-185-0 50 mg
53-181-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-185-0 Supplier Tocris Bioscience™ Supplier No. 5318/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent EGFR and VEGFR inhibitor

AEE 788 is a potent EGFR and VEGFR inhibitor (IC50 values are 2, 6, 59, 77, 160 and 330 nM for EGFR, ErbB2, VEGFR-1, VEGFR-2, ErbB4 and VEGFR-3 respectively). Also inhibits c-Abl, c-Fms and c-Src (IC50 values are 52, 60 and 61 nM respectively). Inhibits proliferation of EGFR and ErbB2 overexpressing cancer cell lines in vitro. Inhibits tumor xenograft growth and VEGF-induced angiogenesis in mice. Orally active.

Specifications

Quantity 50 mg
Product Type AEE 788
Purity >98%
Molecular Weight (g/mol) 440.58
Inhibitors AEE 788

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.