Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Rehydration Fluid for Culti-Loops Organisms, Aerobic Set
Click to view available options
Quantity:
10 Vials/Set
Description
Rehydration fluid set for Thermo Scientific™ Culti-Loops™ aerobic organisms.
Specifications
Specifications
| Description | Aerobic Set Rehydration Fluid |
| Format | Vial |
| Product Type | Quality Control |
| Target Organism Class | Organism Sets |
| Packaging Type | 10 Vial/Set |
| Quantity | 10 Vials/Set |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction