Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aerosol™ OT, Anhydrous Solid, Fisher Chemical

Catalog No. a349500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g

Specific gravity 1.1

  • Anionic waxy solid surfactant for applications where water cannot be tolerated
  • Efficient wetting agent, dispersant, and emulsifier for a variety of applications
  • Used to reduce surface tension and to incrase absorbency and penetration
  • This material is a waxy solid
  • Fisher Chemical also offers a 75% solution in ethanol and water (Catalog No.: A345)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym Dioctyl Sulfosuccinate, AOT, DOSS
PubChem CID 23673837
IUPAC Name sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

Color White
Physical Form Solid
Melting Point 158.9°C
Density 1.1g/cm³
Quantity 500 g
Identification Pass Test
Packaging Poly Bottle
Linear Formula CH3(CH2)3CH(C2H5)CH2O2CCH2CH(SO3Na)CO2CH2CH(C2H5)(CH2)3CH3
Formula Weight 444
Chemical Name or Material Aerosol™ OT, Solid Anhydrous
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause central nervous system effects.
NFPA
Health:
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.