Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AES Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144208 Supplier Invitrogen™ Supplier No. PA5144208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AES Polyclonal Antibody for IHC (P), ELISA

Adhesion to extracellular matrix regulates cell survival through both integrin engagement and appropriate cell spreading. Anoikis is the molecular mechanism of apop-tosis induced by integrin detachment. Amino-terminal enhancer of split (AES) is a member of the Groucho/ transducin-like enhancer of split (TLE) family of transcriptional regulators, a group of transcriptional co-repressors that play important roles in neurogenesis, segmentation, and sex determination. AES forms a complex with Bit1 (Bcl-2 inhibitor of transcription 1), a mitochondrial protein that is released into the cytoplasm upon onset of apoptosis. It has been suggested that this complex turns off a survival-promoting gene transcription program controlled by the TLE protein family. Interestingly, apoptosis of cells transfected with AES and Bit1 could be inhibited if the cells were allowed to attach to fibronectin through the alpha5beta1 integrin suggesting that the Bit1-AES pathway contributing to anoikis is regulated by integrins, and in particular, the alpha5beta1 integrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AES
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 4.235 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TLE5
Gene Accession No. Q08117
Gene Alias Aes; AES-1; AES-2; AL024115; Amino enhancer of split; amino-terminal enhancer of split; Esp1; gp130-associated protein GAM; Grg; Grg5; Grg-5; Groucho-related protein 5; Protein ESP1; Protein GRG; related to Drosophila groucho; R-esp1; TLE family member 5; TLE family member 5, transcriptional modulator; TLE family member 5; amino-terminal enhancer of split; TLE5
Gene Symbols AES, TLE5
Host Species Rabbit
Immunogen Recombinant Human Amino-terminal enhancer of split protein (1-264AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.