Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ Aes Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114846
Description
Antibody detects endogenous levels of total Amino-terminal enhancer of split.
Adhesion to extracellular matrix regulates cell survival through both integrin engagement and appropriate cell spreading. Anoikis is the molecular mechanism of apop-tosis induced by integrin detachment. Amino-terminal enhancer of split (AES) is a member of the Groucho/ transducin-like enhancer of split (TLE) family of transcriptional regulators, a group of transcriptional co-repressors that play important roles in neurogenesis, segmentation, and sex determination. AES forms a complex with Bit1 (Bcl-2 inhibitor of transcription 1), a mitochondrial protein that is released into the cytoplasm upon onset of apoptosis. It has been suggested that this complex turns off a survival-promoting gene transcription program controlled by the TLE protein family. Interestingly, apoptosis of cells transfected with AES and Bit1 could be inhibited if the cells were allowed to attach to fibronectin through the alpha5beta1 integrin suggesting that the Bit1-AES pathway contributing to anoikis is regulated by integrins, and in particular, the alpha5beta1 integrin. 
            Specifications
| Aes | |
| Polyclonal | |
| Unconjugated | |
| TLE5 | |
| Aes; AES-1; AES-2; AL024115; Amino enhancer of split; amino-terminal enhancer of split; Esp1; gp130-associated protein GAM; Grg; Grg5; Grg-5; Groucho-related protein 5; Protein ESP1; Protein GRG; related to Drosophila groucho; R-esp1; TLE family member 5; TLE family member 5, transcriptional modulator; TLE family member 5; amino-terminal enhancer of split; TLE5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14797, 166, 29466 | |
| -20°C | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P63002, P63003, Q08117 | |
| AES, TLE5 | |
| A synthesized peptide derived from human AES(Accession Q08117), corresponding to amino acid residues M1-L43. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                             
                             
                             
                            