Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AF17 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501562022
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2022 Supplier Bethyl Laboratories, Inc Supplier No. A302198A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

MLLT6 gene ontology annotations related to this gene include negative regulation of histone H3-K79 methylation; negative regulation of urine volume; positive regulation of sodium ion transport; positive regulation of transcription by RNA polymerase II; renal potassium excretion; renal sodium excretion.

Specifications

Antigen AF17
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MLLT6
Gene Accession No. P55198
Gene Alias AF17, ALL1-fused gene from chromosome 17 protein, Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 6, protein AF-17, trithorax homolog
Gene Symbols MLLT6
Host Species Rabbit
Immunogen Between 1043 and 1093
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246198, 4302
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.